3-Bromophenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
A367247-1G
A367247-25G
A367247-5G
A367247-100G
A367247-10G
A367247-500G
103826-422EA
7
USD
103826-422
103837-432
76840-664
76840-666
76840-668
77115-362
77115-364
77115-366
3-Bromophenylboronic acid 97%
3-Bromophenylboronic acid
Caution: for Research Use Only
Formula:
C₆H₆BBrO₂ Boiling Pt: 330.5 °C (760 mm Hg) Melting Pt: 164…168 °C Flash Pt: 153.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00239386 CAS Number: 89598-96-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...