((3-Bromopropoxy)methyl)benzene 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzyl 3-bromopropyl ether, 3-bromopropylbenzyl ether
A319797-100G
A319797-10G
A319797-1G
A319797-25G
A319797-5G
103825-218EA
7
USD
103825-218
103836-150
76772-906
76772-908
76839-000
76839-002
76839-004
((3-Bromopropoxy)methyl)benzene 96%
((3-Bromopropoxy)methyl)benzene
Caution: for Research Use Only
Formula:
C₁₀H₁₃BrO MW: 229.117 g/mol Boiling Pt: 251.7±15.0 °C (760 mm Hg) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134570 CAS Number: 54314-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...