NEBExpress Ni Spin Columns, New England Biolabs
Supplier: New England Biolabs (NEB)
S1427L S1427S
76377-482EA 239 USD
76377-482 76377-484
NEBExpress Ni Spin Columns, New England Biolabs
Filters Centrifugal Filters
Includes ready-to-use pre-packed Ni spin columns and all required buffers for purification.
  • Purification of ≥1 mg His-tagged protein per column in as little as 15 min
  • High specific binding of His-tagged proteins yielding purities of >95% Strong nickel ion binding provides excellent resistance to EDTA and reducing agents
  • Compatible with isolation and purification of His-tagged fusion proteins under native or denaturing conditions