AleI-v2
Supplier: New England Biolabs (NEB)
R0685L R0685S
76326-054EA 331 USD
76326-054 76326-056
AleI-v2
Nucleic Acid Reagents Restriction Enzymes
AleI-v2 is an engineered and improved version of AleI that recognizes the sequence CACNN^NNGTG.
  • Cut site: CACNN/NNGTG
Concentration: 10000 units/ml.