Esp3I
Supplier: New England Biolabs (NEB)
R0734L R0734S
76284-688EA 402 USD
76284-688 76284-690
Esp3I
Enzymes
A Type IIS restriction endonuclease that recognizes the sequence CGTCTCN^NNNN_....
  • Time-Saver qualified for digestion in 5-15 minutes
  • Isoschizomer of BsmBI-v2
  • Used in Golden Gate AssemblyType IIS REs recognize asymmetric DNA sequences; cleave outside their recognition sequence
  • Supplied with 1 vial of Gel Loading Dye, Purple (6X)