Western Blotting Mouse IgG DAB Chromogenic Reagent Kit (Yellow), Boster
Supplier: Avantor
76468-106EA 106.24 USD
76468-106
Western Blotting Mouse IgG DAB Chromogenic Reagent Kit (Yellow), Boster
Blotting Kits
All-In-One kit with HRP conjugated secondary antibody and DAB substrate, optimized for Western Blotting.