Human IL-22 ELISA Kit EZ-Set (DIY Antibody Pairs), Boster
Supplier: Boster Biological Technology
EZ0933
76467-242EA 546.34 USD
76467-242
Human IL-22 ELISA Kit EZ-Set (DIY Antibody Pairs), Boster
Assays ELISAs
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects IL-22 in Human samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC.