nBIONIX Hypoxic Cell Culture Kit, Bulldog Bio
Supplier: Bulldog Bio
NBIONIX-3
103881-422EA 1615.38 USD
103881-422
nBIONIX Hypoxic Cell Culture Kit, Bulldog Bio
Cell Culture Media
The nBionix Hypoxid Cell Culture Kit offers every lab an easy, adaptable and inexpensive hypoxia 'chamber'.
The nBIONIX Hypoxic Cell Culture Kit enable convenient, precise and inexpensive hypoxic cultivation. It can take less than 10 minutes to create a precisely tuned low oxygen environment from 0.1 to 15% O₂ levels. The gas-barrier pouch contains a precision O₂ meter (to provide continuous feedback), in addition to one or more dishes or plates of cells. This easily transportable micro-environment can be placed in a standard cell culture incubator for long-term incubations under hypoxic conditions.