Anti-DNP HEK293 Polyclonal Antibody(FITC Labeled Protein)

Supplier: ACRO Biosystems
DNPFM2200UG DNPFM225UG
103870-880EA 798 USD
103870-880 103870-882
Anti-DNP HEK293 Polyclonal Antibody(FITC Labeled Protein)
Antibodies
Type: Primary
Antigen: DNP
Clonality: Polyclonal
Clone:
Conjugation: FITC Labeled Protein
Epitope:
Host: HEK293
Isotype: Human IgG1/kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR