Anti-Adalimumab Hybridoma Polyclonal Antibody

Supplier: ACRO Biosystems
ADBY191MG ADBY19100UG
103815-448EA 3234 USD
103815-448 103815-450
Anti-Adalimumab Hybridoma Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Adalimumab
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hybridoma
Isotype: IgG1/kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR