Anti-Bevacizumab Hybridoma Polyclonal Antibody (For ADA assay)

Supplier: ACRO Biosystems
BEBY91MG BEBY9100UG
103815-434EA 3234 USD
103815-434 103815-436
Anti-Bevacizumab Hybridoma Polyclonal Antibody (For ADA assay)
Antibodies
Type: Primary
Antigen: Bevacizumab
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hybridoma
Isotype: IgG1/kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR