Anti-Cetuximab Hybridoma Polyclonal Antibody (Non-Neutralizing)

Supplier: ACRO Biosystems
CEBY311MG CEBY31100UG
103815-422EA 3234 USD
103815-422 103815-424
Anti-Cetuximab Hybridoma Polyclonal Antibody (Non-Neutralizing)
Antibodies
Type: Primary
Antigen: Cetuximab
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hybridoma
Isotype: IgG1/kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR