Sign, 'DANGER CONTROLLED AREA, CLOSED FOR DECONTAMINATION' Accuform®
Supplier: Accuform
MBHZ134VA MBHZ136VP MBHZ134VP MBHZ136VS MBHZ134VS MBHZ136VA
76441-764EA 28.18 USD
76441-764 76441-762 76441-768 76441-766 76441-772 76441-770
Sign, 'DANGER CONTROLLED AREA, CLOSED FOR DECONTAMINATION' Accuform®
Signs
Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage.
  • Multiple material and size options
Choose from three material options. Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. Moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. Plastic signs are durable with slight flex. Good material choice for indoor and sheltered outdoor environments. The printed surface has a clear UV protective overcoat. Aluminum signs provide the strength of metal without rusting. Printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance.