Edvotek BactoBeads™ Lyophilized Bacteria
Supplier: Edvotek
741 729 728 726 743 161-BC 731 953-252 160BACTOBEADS
470331-000EA 235.1 USD
470331-000 470331-010 470331-004 470331-002 470330-999 470331-012 470331-008 470331-018 470331-006
Edvotek BactoBeads™ Lyophilized Bacteria
Educational Materials Biology Educational Materials Microbiology Learning Activities
Place one BactoBead™ on an agar plate, watch it dissolve, and streak for isolated colonies.
  • Quick and convenient
  • Non-pathogenic lyophilized bacteria
BactoBeads™ absorb moisture quickly so keep the vial closed and refrigerated when not in-use. Reconstitute only what you need when you need it.
Store vial containing unused beads in refrigerator in a zip-top bag with desiccant.
Ordering information: Items ship immediately upon ordering. Please call to specify a future shipping date.
Caution: Do not store in a refrigerator with food for human or animal consumption. Follow all safety guidelines in the package.