Phthalates in Polyvinyl Chloride Standard, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
CRM-PVC001 CRM-PVC003
SXCRM-PVC001EA 152.93 USD
SXCRM-PVC001 SXCRM-PVC003
Phthalates in Polyvinyl Chloride Standard, SPEX CertiPrep
Standards Environmental Analysis Standards
Consumer safety standards, phthalates in polyvinyl chloride (PVC) QC standards.
Polyvinyl chloride, or PVC, is a very common plastic used in a wide range of common consumer products from children’s toys and care items to building and construction materials. In the US, ASTM (American Society for Testing and Materials) and CPSC (Consumer Product Safety Commission) have designed methods for testing children’s toys and childcare articles for compliance with the restriction of use for eight designated phthalates.