Bacitracin

Supplier: RPI

B32000-1.0 B32000-25.0 B32000-5.0 B32000-100.0
76345-128EA 18.69 USD
76345-128 76345-130 76345-132 77510-186
Bacitracin
Bacitracin

Interferes with bacterial cell wall synthesis.


  • Active against gram positive bacteria
  • Toxic to plant cells


Caution: Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₆₆H₁₀₃N₁₇O₁₆S₁
Melting Pt: 221…225 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062640
CAS Number: 1405-87-4
EINECS: 215-786-2

Order Now

Specification Test Results

Appearance: White to pale buff powder
pH: 5.5 - 7.5
Loss on Drying: ≤ 5.0 %
Residue on Ignition: ≤ 0.5 %
Source: Bacillus Licheniformis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR