Alpha-Synuclein Rapid ELISA Kits: Human, Biosensis
Supplier: Biosensis
The Biosensis Alpha-Synuclein Rapid enzyme-linked immunosorbent assay (ELISA) kit is a sandwich ELISA that allows the quantification of human alpha-synuclein in less than 4 hours in human citrate-plasma, serum and CSF only if used as directed.
- Complete kit with precise instructions
- Assay duration of less than 4 hours without compromising quality
- Kit has been validated for accurate quantification of human Alpha-Synuclein in blood and CSF
This ELISA kit consists of a pre-coated sheep polyclonal anti-alpha-synuclein (aa: 116-131) capture antibody, a biotinylated mouse monoclonal anti-alpha-synuclein detection antibody (aa: 61-95) and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3',5,5'-tetramethylbenzidine, TMB) yields a colored reaction product which is directly proportional to the concentration of alpha-synuclein present in samples and protein standards. A human alpha-synuclein positive control (QC sample) is provided to assure consistent assay performance. Please refer to the kit protocol for specific use instructions for blood and CSF application.
Specificity: Human alpha-synuclein. Detection of individual alpha-synuclein isoforms has not yet been investigated. However, due to the nature of the assay antibodies, this ELISA kit is not expected to discriminate between various alpha-synuclein isoforms (monomeric, oligomeric, phosphorylated). Cross-reactivity with beta- and gamma-synuclein was calculated based on increase or decrease of apparent endogenous α-synuclein concentrations when spiking beta- and gamma-synuclein into two human plasma samples. This ELISA shows very little cross-reactivity of 2.3% or less for beta- and gamma-synuclein.
Delivery information: The ELISA kit box contains 1×96-well pre-coated strip plate, protein standards, QC sample, detection reagents, wash and sample buffers, substrate buffer and detailed protocols.
Caution: This ELISA kit has not been tested for other applications. It has been configured for research use only and is not to be used for diagnostic or clinical procedures.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...