Acrylamide : Bisacrylamide (37.5:1), powder, Premixed

Supplier: RPI
Danger

A11310-100.0 A11310-500.0
76344-558EA 120.88 USD
76344-558 76344-560
Acrylamide : Bisacrylamide (37.5:1), powder, Premixed
Acrylamide-Bis (32:1)

Acrylamide/bis-Acrylamide, Premixed Powder 37.5:1 Ratio


  • Pre-measured powder blend of acrylamide and bis-acrylamide for simple stock solution preparation.

Storage Temperature: Ambient
MDL Number: MFCD00080848
UN: 2074
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White powder
Conductivity of Solution ≤ 10 µmhos
Electrophoresis Pass Test
Solubility Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR