VersaPlate 96-Well-PPL
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
75480060 75409025
101473-748EA 484.23 USD
101473-748 103579-942 103579-946 103579-948 103579-952 103579-954 AG75480060 AG75409025
VersaPlate 96-Well-PPL
Microplates Solid Phase Extraction Plates (SPE)
VersaPlate allows you to process more samples in less time with less waste in a 96-well format.
  • Use only the number of wells you need for each run to save on sample preparation costs, while still enjoying all the benefits of automation
  • Increased well depth reduces the risk of overflow, and flangeless, beveled rims provide a tight fit. This helps avoid cross contamination
  • Scale up using the exact same product that was used with small scale development
Develop an SPE method faster, using liquid handling automation or multichannel pipettes, or use individual tubes to quickly work out method conditions and transfer to 96-well processing.
The Agilent VersaPlate allows you to process more samples in less time with less waste. In its 96-well format, tubes are removable, making this system easier to implement, simpler to use, and far more flexible than conventional 96-well formats. Whether you need to speed the processing of hundreds of samples, validate just a few samples, or develop a new method, you can customize the configuration to meet your needs.