Anti-IgG Rabbit Polyclonal Antibody (Gold, 10nm)

Supplier: Electron Microscopy Sciences
25228 25229
100487-602EA 645.41 USD
100487-602 100487-604
Anti-IgG Rabbit Polyclonal Antibody (Gold, 10nm)
Antibodies
The Conventional Immunogold Reagents are the classical conjugates in immuno electron microscopy; they are a good choice when the antigen is abundant and the accessibility of the antigen is relatively good.

Conventional ImmunoGold Reagents are monodisperse size population makes the conjugates suited for multiple labeling with no overlap. The Conventional Immunogold Reagents are the classical conjugates in immuno electron microscopy; they are a good choice when the antigen is abundant and the accessibility of the antigen is relatively good. The conventional labeling approach in transmission and scanning electron microscopy utilizes secondary immunogold reagents based on particles that can be observed without enhancement. The particle population is monodisperse and thus shows minimal size variation and overlap.The reagents are supplied in PBS with 1% Bovine Serum Albumin and 15 mM NaN3.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Gold, 10nm
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR