The IMEB Bone Band Saw is an efficient and economical bone cutting tool for the laboratory. The lightweight, portable device is only 19" high and occupies a footprint of 17×13" of counter space.
- Efficient and economical tool for cutting bone
- Saw is constructed of aluminum and stainless steel for durability
- Saw will cut bone specimens up to 3" in height
- Lightweight and easy to clean
- Portable, small table-top unit measures 17×13×19"
- Strong blade can cut ultra-thin bone slices
- Blade guide is easily adjusted
A strong blade is included that can cut ultra-thin bone slices. An optional water-cooled Diamond blade reduces dust and particles. Four blade types and additional accessories are available to provide a versatile laboratory bone cutting station.
Accessories information: Optional accessories include four blade types, cooling systems, femoral and small bone holders, splash guards, cleaning stones, and histo-vacuum. The optional cooling system is required for use with Diamond saw blades.
Delivery information: Standard blade, mounting board, edge guide, and foot switch included.