30 - Tablet or Chromebook Charging Cart, Luxor
Supplier: Luxor
LLTM30-B
76203-828EA 1853.36 USD
76203-828
30 - Tablet or Chromebook Charging Cart, Luxor
Carts
Luxor tablet or chromebook charging carts safely charge, store and transport tablets and chromebooks to meet the evolving IT needs of classrooms and dynamic work environments.
  • Rubber-coated dividers safeguard devices from damage
  • Bottom storage shelf for storing additional IT items
  • Locking front and rear access doors; includes two keys
  • All-steel construction and padded top surface
  • Four non-marring 4" casters, two with locking brakes, and handle for easy maneuverability
  • Ventilated on all four sides, allowing air to circulate freely through the unit
  • Side cord wrap for convenience
  • Includes two UL-Listed 15-outlet 120 V/15 A vertical electrical power strips
  • Number of cords: two
A strong, simple design with lockable steel doors provides rugged security and storage while inside, an electrical assembly and cord management system keep things organized and charged up. Rubber-coated dividers won't scratch devices and help provide impact protection during transit, which is made easy even when fully loaded thanks to rolling casters and a full-length handle. And the cabinet requires minimal assembly – simply attach the casters and handle.