Non-Adhesive Formalin-Resistant Tags
Supplier: GA INTERNATIONAL INC
807656026800 807656026817 807656026824 807656026831 807656026848 807656026855
76276-178RL 48.57 USD
76276-178 76276-180 76276-182 76276-184 76276-186 76276-188
Non-Adhesive Formalin-Resistant Tags
Labels
Thermal transfer printable tags for the long-term identification of specimens preserved in 10 to 100% formalin and alcohol solutions. These non-adhesive tags are resistant to formaldehyde and alcohol, and are intended to be inserted directly into the container with preserved specimens.
  • Temperature range: −196 to +150 °C (−321 to +302 °F)
  • Able to withstand long-term immersion in harsh chemicals and specimen liquid preservatives
  • Supplied in a continuous roll for ease of use and size selection
  • Printable with all major thermal transfer printer brands
Chemical-resistant, non-adhesive tags designed for storage in specimen jars and containers for the long-term identification of samples (i.e., animals, tissues, organs) in preservatives including formalin, alcohol, as well as formalin-acetic acid-alcohol combinations. Made of a chemically inert material, they will not affect specimen integrity. Supplied in continuous roll format and compatible with all major brands of thermal transfer printers with cutter function to select the desired length. When printed with our recommended chemical-resistant ribbons, the printout is also resistant to long-term storage in liquid preservatives.