KIMBLE® Liebig Condensers with Standard Taper Joints, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
447000-2430 447000-2410 447000-2420 447000-2425 447000-2440
KT447000-2430EA 306.8 USD
KT447000-2430 KT447000-2410 KT447000-2420 KT447000-2425 KT447000-2440 KT447000-2920
KIMBLE® Liebig Condensers with Standard Taper Joints, DWK Life Sciences
Condensers
These Liebig condensers have a standard taper outer joint at the top and a inner drip joint at the bottom.
  • Hose connections accept ³/₈" ID flexible tubing
  • Ref: ASTM Method D322
  • Manufactured from 33 expansion, low extractable borosilicate glass
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.