Petroleum spirit, 60…80 °C, Puriss. p.a., high boiling, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
32248-1L
32248-2.5L
BJ32248-1LEA
113.07
USD
BJ32248-1L
BJ32248-2.5L
Petroleum spirit, 60…80 °C, Puriss. p.a., high boiling, Riedel-de Haën™
Petroleum spirit
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Boiling Pt:
63…80 °C (1.013 hPa) Density: 0.68 g/cm³ (20 °C) Flash Pt: 7.4 %(V) Storage Temperature: Ambient |
MDL Number:
MFCD00081849 CAS Number: 64742-49-0 EINECS: 309-852-0 UN: 3295 ADR: 3,II |
Specification Test Results
Boiling range | 60 - 80 °C |
Density (D 20/4) | 0.660 - 0.680 |
Refractive index (n 20/D) | 1.376 - 1.382 |
Non-volatile matter | Max. 0.001 % |
Water (Karl Fischer) | Max. 0.01 % |
Free acid (as CH₃COOH) | Max. 0.002 % |
Aluminium (Al) | Max. 0.00005 % |
Boron (B) | Max. 0.000002 % |
Barium (Ba) | Max. 0.00001 % |
Calcium (Ca) | Max. 0.00005 % |
Cadmium (Cd) | Max. 0.000005 % |
Cobalt (Co) | Max. 0.000002 % |
Chromium (Cr) | Max. 0.000002 % |
Copper (Cu) | Max. 0.000002 % |
Iron (Fe) | Max. 0.00001 % |
Magnesium (Mg) | Max. 0.00001 % |
Manganese (Mn) | Max. 0.000002 % |
Nickel (Ni) | Max. 0.000002 % |
Lead (Pb) | Max. 0.000002 % |
Tin (Sn) | Max. 0.00001 % |
Zinc (Zn) | Max. 0.00001 % |
Benzene (GC) | Max. 0.005 % |
Iodine value | Max. 0.3 |
S-compounds (as S) | Max. 0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...