Ethyl acetate ≥99.0% (by GC), Laboratory Reagent
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Acetic acid ethyl ester, Acetic ether, Acetoxyethane, Acetidin, Ethyl acetic ester, Ethyl ethanoate, EtOAc, Vinegar naphtha, Acetic acid ethyl ester
16371-4X5L
16371-5L
BJ16371-4X5LCS
871.9
USD
BJ16371-4X5L
BJ16371-5L
Ethyl acetate ≥99.0% (by GC), Laboratory Reagent
Ethyl acetate
- ≥95% for general laboratory use.
Beilstein Number: 506104
RTECS Number: AH5425000
UNSPSC Code: 12191602
Formula:
CH₃COOC₂H₅ MW: 88.11 g/mol Boiling Pt: 77.1 Melting Pt: -83 Density: 0.901 g/cm³ Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009171 CAS Number: 141-78-6 EINECS: 205-500-4 UN: 1173 ADR: 3 II Merck Index: 13,03792 |
|
Specification Test Results
Assay (GC) | Min. 99.0 % |
Free acid (as CH₃COOH) | Max. 0.003 % |
Water (Karl Fischer) | Max. 0.05 % |
Non-volatile matter | Max. 0.003 % |
Explosion Limit | 2.2-11.5 %, 38 °F |
Refractive Index | n20/D 1.3720 (lit.) |
Autoignition Temperature | 801 °F |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...