FastAmp® Plant Direct PCR Kit, Intact Genomics
Supplier: INTACT GENOMICS, INC MS
FastAmp™ plant direct PCR kit is suitable for amplification of DNA directly from plant samples without purifying DNA.
- Direct PCR- no need to purify DNA
- Specially engineered Taq DNA polymerase with highest sensitivity and specificity
- Short PCR protocol times
- Master mix format with premixed gel loading dye to reduce cross-contamination and sample handling errors
- 5X magic enhancerfor high GC containing DNA amplification
- Quality control is performed following the production of each new lot of product to ensure that it meets the quality standards and specifications designated for the product
- FastAmp™ plant direct PCR master mix (2X)
- Dilution buffer
- Control primer mix (25 µM each)
- Nuclease- free water
FastAmp™ plant direct PCR kit is an easy and robust method to amplify DNA fragments from plant tissues with high specificity and high sensitivity without the need of complicated DNA purification steps. The advanced formulation of this kit also allows fast PCR cycling conditions without compromising PCR sensitivity, specificity and yield. The FastAmp™ plant direct PCR kit is an ideal and powerful tool for high-throughput genotyping, DNA amplification, and plant genome analysis.
FastAmp™ plant direct PCR kit is suitable for amplification of DNA directly from plant samples without purifying DNA. This kit is based on specially engineered Taq DNA polymerase, proprietary buffer system, dNTP, MgCl₂, PCR facilitators and dye mix which makes it extremely robust and tolerant of plant PCR inhibitors such as complex polysaccharides, polyphenols and others. This kit has been tested with leaves and seeds from a wide variety of plant species.
Delivery information: FastAmp™ plant direct PCR kit includes a complete set of optimized reagents and detailed protocols making it an ideal choice for amplification of plant DNA without DNA purification. All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (central time, monday-thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on friday will be ship on the following monday. The shipping and dry ice packaging cost is $40 for your entire order in the 48 contiguous United States.