Human Recombinant MCP-2 (CCL8) (from E. coli)
Supplier: Peprotech
AF-300-15-2UG AF-300-15-10UG AF-300-15-50UG AF-300-15-100UG AF-300-15-250UG AF-300-15-500UG AF-300-15-1MG
76304-160EA 158.05 USD
76304-160 76304-162 76304-164 76304-166 76304-234 76304-236 76304-238
Human Recombinant MCP-2 (CCL8) (from E. coli)
Proteins and Peptides
The MCP proteins are members of the CC chemokine family that signal through CCR2 and, with the exception of MCP-1, other CCR receptors. The MCP proteins chemoattract and activate monocytes, activated T cells, basophils, NK cells, and immature dendritic cells. The MCP family cross-reacts across species. Recombinant Human MCP-2 (CCL8) is an 8.9 kDa protein containing 76 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines.
  • Animal-free