Anti-IgG Rabbit Polyclonal Antibody Amphiregulin

Supplier: Peprotech
500-P322-50UG 500-P322-100UG 500-P322-1MG
76304-308EA 317.82 USD
76304-308 76304-310 76304-312
Anti-IgG Rabbit Polyclonal Antibody Amphiregulin
Antibodies
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR