Anti-IgG Rabbit Polyclonal Antibody (Biotin) VEGF-B

Supplier: Peprotech
500-P267BT-25UG 500-P267BT-50UG
76304-114EA 317.82 USD
76304-114 76304-116 76304-118
Anti-IgG Rabbit Polyclonal Antibody (Biotin) VEGF-B
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR