Anti-IgG Rabbit Polyclonal Antibody CRP

Supplier: Peprotech
500-P242-50UG 500-P242-100UG 500-P242-1MG
76304-292EA 317.82 USD
76304-292 76304-294 76304-296
Anti-IgG Rabbit Polyclonal Antibody CRP
Antibodies
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR