Anti-IgG Rabbit Polyclonal Antibody sIL-4 Receptor alpha

Supplier: Peprotech
500-P325-50UG 500-P325-100UG 500-P325-1MG
76304-186EA 317.82 USD
76304-186 76304-188 76304-190
Anti-IgG Rabbit Polyclonal Antibody sIL-4 Receptor alpha
Antibodies
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR