Anti-IgG Rabbit Polyclonal Antibody (Biotin) p16-INK-4a TAT

Supplier: Peprotech
500-P284TBT-25UG 500-P284TBT-50UG
76304-006EA 317.82 USD
76304-006 76304-008 76304-010
Anti-IgG Rabbit Polyclonal Antibody (Biotin) p16-INK-4a TAT
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR