Anti-IgG Rabbit Polyclonal Antibody Lymphotactin (XCL1)

Supplier: Peprotech
500-P40-50UG 500-P40-100UG
10775-266EA 339.8 USD
10775-266 10775-268
Anti-IgG Rabbit Polyclonal Antibody Lymphotactin (XCL1)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human Lymphotactin (XCL1). Anti-Human Lymphotactin (XCL1)-specific antibody was purified by affinity chromatography employing an immobilized Human Lymphotactin (XCL1) matrix.

Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR