L-Tyrosine disodium salt dihydrate ≥99% (by HPLC), J.T.Baker®

Warning

Synonyms: (S-3-(4-Hydroxyphenyl)alanine disodium salt dihydrate, Disodium 2-Amino-3-(4-oxidophenyl) propanoate dihydrate, L-3-(4-Hydroxyphenyl)alanine disodium salt dihydrate

Management of Change (MOC) category = R

BS23-SA
JTBS23-SACS 851.09 USD
JTBS23-SA
L-Tyrosine disodium salt dihydrate ≥99% (by HPLC), J.T.Baker®
L(-)-Tyrosine disodium salt
Formula: C₉H₉NO₃Na₂·2H₂O
MW: 261.19 g/mol
MDL Number: MFCD03840558
CAS Number: 122666-87-9

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Appearance (White to beige powder with occasional brown solid) Passes Test
Identification (IR) Passes Test
Solution (5%) in water Passes Test
Assay (by HPLC) ≥99 %
Water (by KF) ≤19.3 %
Bacterial endotoxins, EU/g ≤50
Aluminum (Al)
Iron (Fe)
Cadmium (Cd)
Cobalt (Co)
Nickel (Ni)
Vanadium (V)
Selenium (Se)
Chromium (Cr)
Copper (Cu)
Manganese (Mn)
Molybdenum (Mo)
Tin (Sn)
Zinc (Zn)
Total Aerobic Microbial Count ≤100 cfu/g
Total Yeasts and Molds ≤100 cfu/g
Residual Solvents: Class 3, each ≤5000 ppm
Residual Solvents: Only Class 3 solvents are present as a result of use
in the purification process. The concentration of Class 3 solvents is
determined and reported for each production batch.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR