Irradiated Contact Plates Media, BioMerieux
Supplier: bioMerieux
These contact plates can be used for surface environmental monitoring.
Irradiated Count-Tact 3P agar is used for the monitoring of microbial contamination in industrial and hospital clean room environments.
Dey-Engley agar is a medium used for the detection and enumeration of micro-organisms that are present in environments that contain antimicrobial chemicals, such as surfaces that are of sanitary importance.
Irradiated Tryptic Soy Agar (TSA) with Lecithin and Polysorbate 80 is used for the quantitation of viable microbial contaminants from surfaces and equipment in controlled environments.
Sabouraud Dextrose Agar with Lecithin and Polysorbate 80 is for isolation and cultivation of fungi from surfaces sanitized with quaternary ammonium compounds.
Ordering information: Made to order items have a lead time of 42 days.