Simmons Citrate Agar
Supplier: VWR International
AD-MM1040-100G
AD-MM1040-500G
470317-446EA
88
USD
470317-446
470317-448
Simmons Citrate Agar
Media for Microbiology
Prepared Media for Microbiology
A medium helpful in differentiation of the enterobacteriaceae. Simmons citrate agar test for organisms that are able to utilize citrate as a carbon source.
To prepare media, add 24 g of Simmons citrate agar to a liter of distilled or de-ionized water. The media will not be sterile until autoclaved prior to use.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)