Sim Medium
Supplier: VWR International
AD-MM1041-100G
AD-MM1041-500G
470317-450EA
49.45
USD
470317-450
470317-452
Sim Medium
Media for Microbiology
Dehydrated Media for Microbiology
To assist in the differentiation of enteric organisms the medium provides three tests - indole production, production of hydrogen sulfide by organisms that can reduce sulfur, and motility.
- Requires Kovac’s reagent for determination of indole production
To prepare media, add 30 g of SIM medium to a liter of distilled or de-ionized water. The media will not be sterile until autoclaved prior to use.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)