Somso® Mitosis and Meiosis
Supplier: MARCUS SOMMER
ZOS 57/3 ZOS 57/1
470105-088KT 1162.39 USD
470105-088 470102-816
Somso® Mitosis and Meiosis
Models Human Biology Models
Somso® premium quality models.
  • 5 year warranty
Enlarged many times, in SOMSO-Plast®. After Christian Groß, Head of Biology Department (retired). In 8 separate models the following phases are represented: 1. late interphase, 2. prophase, 3. beginning of metaphase, 4. metaphase - equatorial plane, 5. anaphase - migration of daughter chromosomes, 6. late anaphase, 7. telophase and 8. daughter cells in the early interphase. As shown in the models 3 - 5 the spindles and chromosomes can be revolved for demonstration purposes under a transparent cover. In one piece. The models are mounted individually on a stand with green base.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.