Zein
Supplier: Spectrum Chemical Mfg. Corp.
Zein, a class of prolamine protein, is found in corn and used in many commercial products including clothing fabric, paper cups, adhesives, and textile fibers.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
MDL Number:
MFCD00062414 CAS Number: 9010-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...