Xanthane hydride

Supplier: Spectrum Chemical Mfg. Corp.

X1072-100GM X1072-25GM
75873-566EA 434.2 USD
75873-566 75873-568
Xanthane hydride
Xanthane hydride
Formula: C₂H₂N₂S₃
Storage Temperature: Ambient
MDL Number: MFCD00051660
CAS Number: 6846-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR