1-Pentanesulfonic acid sodium salt, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate, Pentane-1-sulfonic acid sodium salt, Sodium pentanesulfonate, Sodium 1-pentanesulfonate, Sodium pentane-1-sulfonate, Sodium pentanesulfonate, 98%

S2750-100GM S2750-25GM S2750-500GM
75873-340EA 309.7 USD
75873-340 75873-342 75873-344
1-Pentanesulfonic acid sodium salt, Reagent Grade
1-Pentanesulphonic acid sodium salt
Formula: C₅H₁₁O₃SNa
MW: 174.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS Number: 22767-49-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddb1c23c8a6fe3c156a3aacb62721e1d1a34efb0&vid=1936dcdb47ba374ce21fd758660f7ca6ba95d254&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR