Pyrazine-2-carboxylic acid

Supplier: Spectrum Chemical Mfg. Corp.

P2329-100GM P2329-25GM P2329-5GM
75873-228EA 211.65 USD
75873-228 75873-230 75873-232
Pyrazine-2-carboxylic acid
Pyrazine-2-carboxylic acid
Formula: C₅H₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00006130
CAS Number: 98-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR