5-Nitro-o-toluidine

Supplier: Spectrum Chemical Mfg. Corp.

M2823-100GM M2823-25GM
75872-434EA 299.1 USD
75872-434 75873-024
5-Nitro-o-toluidine
5-Nitro-o-toluidine
Formula: C₇H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00007741
CAS Number: 99-55-8
UN: 2660
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR