3-Methyl-2-benzothiazolinonehydrazone hydrochloride hydrate

Supplier: Spectrum Chemical Mfg. Corp.

M2140-25GM M2140-5GM
75873-008EA 278.44 USD
75873-008 75873-010
3-Methyl-2-benzothiazolinonehydrazone hydrochloride hydrate
3-Methyl-2-benzothiazolinonehydrazone hydrochloride hydrate
Formula: C₈H₉N₃S
Storage Temperature: Ambient
MDL Number: MFCD00149370
CAS Number: 149022-15-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR