(±)-α-Lipoic acid
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)-1,2-Dithiolane-3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid
alpha-Lipoic Acid is also known as alpha lipoic acid or thioctic acid. It is an organosulfur compound that comes from octanoic acid. It is made most often in animals and is crucial for aerobic metabolism.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₈H₁₄O₂S₂ MW: 206.33 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00005474 CAS Number: 1077-28-7 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...