Inosine 5'-monophosphate disodium salt hydrate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 5'-IMP 2Na hydrate, 5'-Inosinic acid disodium salt hydrate, Inosinic acid (disodium)(hydrate)(1:2:X), Inosine 5'-monophosphate disodium salt xhydrate, Disodium inosinate hydrate, Inosic acid disodium salt hydrate

I3041-100GM I3041-10KGBL I3041-25GM I3041-5GM
75872-814EA 1261.75 USD
75872-814 75872-816 75872-818 75872-820
Inosine 5'-monophosphate disodium salt hydrate
Inosine 5'-monophosphate disodium salt hydrate
Formula: C₁₀H₁₁N₄Na₂O₈P·xH₂O
MW: 392.17 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD00036201
CAS Number: 352195-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR