L(+)-Histidine monohydrochloride monohydrate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: L-Histidine monohydrochloride monohydrate, (2S)-(+)-Histidine monohydrochloride monohydrate, L-Histidine hydrochloride monohydrate, (S)-(+)-Histidine monohydrochloride monohydrate, His HCl monohydrate, H-His-OH.HCl.H2O, H-His-OH·HCl·H₂O, L-3-Imidazol-4-ylalanine monohydrochloride monohydrate, L-Histidine hydrochloride hydrate

HI125-5KG HI125-100GM HI125-1KG HI125-25GM
75872-726EA 2789.7 USD
75872-726 75872-744 75872-746 75872-748
L(+)-Histidine monohydrochloride monohydrate
L(+)-Histidine monohydrochloride monohydrate

L-Histidine Monohydrochloride, Monohydrate, also known as 2-Amino-3-(1H-imidazol-4-yl)propanoic acid, is an alpha-amino acid with an imidazole functional group.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₉N₃O₂·HCl·H₂O
MW: 209.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151027
CAS Number: 5934-29-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=561d2381f930afe618f78d680790ea9a43f6596c&vid=8aacf6dc4ef5382410cb743498272b48c3c36999&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR