7-(2-Hydroxyethyl)theophylline
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
7-(2-Hydroxyethyl)-1,3-dimethyl-3,7-dihydro-1H-purine-2,6-dione, Etofylline
H3275-100GM
H3275-25GM
75872-700EA
264.6
USD
75872-700
75872-702
7-(2-Hydroxyethyl)theophylline
7-(2-Hydroxyethyl)theophylline
Formula:
C₉H₁₂N₄O₃ MW: 224.216 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00055055 CAS Number: 519-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...