DL-Glyceraldehyde

Supplier: Spectrum Chemical Mfg. Corp.

G3307-25GM G3307-5GM
75872-556EA 132.82 USD
75872-556 75872-558 75872-560
DL-Glyceraldehyde
DL-Glyceraldehyde
Formula: C₃H₆O₃
Storage Temperature: Ambient
MDL Number: MFCD00064379
CAS Number: 56-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR